1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%
1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%
Thermo Scientific Chemicals

1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043496.14
also known as 043496-14
Price (USD)/ Each
77.70
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
77.70
Add to cart
1,3-Bis[tris(hydroxymethyl)methylamino]propane, 98+%
Catalog number043496.14
Price (USD)/ Each
77.70
-
Add to cart
Chemical Identifiers
CAS64431-96-5
IUPAC Name2-[(3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC11H26N2O6
InChI KeyHHKZCCWKTZRCCL-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCCCNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder
Loss on Drying≤1.0%
Optical Absorbance at400nm: ≤0.05 AU ( 0.1M aqueous solution)
Identification (FTIR)Conforms
Optical Absorbance at280nm:≤0.15( 0.1M aqueous solution)
View more
1,3-Bis[tris(hydroxymethyl)methylamino]propane is used as a buffer for polymerase chain reaction and molecular biology. It is also used with hydrochloric acid buffer for stabilization of farnesyl diphosphate isolated from a strain of saccharomyces cerevisiae. It acts as a ligand and forms dinuclear hydroxo complexes with Lanthanides(III). Further, it is useful for calibration of genetically encoded pH indicators expressed in the cytosol or mitochondria. It is also employed to investigate the influence of anions with varying nucleophilic properties on the autoxidation of oxymyoglobin. In addition to this, it is used in the purification of glucose-binding protein from membranes of sulfolobus solfataricus and as a non-zwitterionic buffer, which is used in the pH range between 6.3 and 9.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only