Thermo Scientific Chemicals

Glycine, ACS, 98.5+%

Catalog number: 043497.36
500 g, Each
Thermo Scientific Chemicals

Glycine, ACS, 98.5+%

Catalog number: 043497.36
500 g, Each
Quantity
Catalog number: 043497.36
also known as 043497-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-40-6
IUPAC Name
2-aminoacetic acid
Molecular Formula
C2H5NO2
InChI Key
DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES
NCC(O)=O
Impurity content
Hydrolyzable substances P.T.
Impurity content
SO₄ 0.005%
Impurity content
Cl 0.005%
Impurity content
NH₄ 0.005%
Trace Metals
(as Pb) 0.002%

Description

Glycine is employed to protect the kidneys from the harmful side effects of certain drugs used after organ transplantation as well as the liver from the harmful effects of alcohol. The solution of glycine plays an important role for deposition of alloys and metals. It is likewise useful for patients with acute alcohol hallucinosis. It is applied for treating schizophrenia, stroke, benign prostatic hyperplasia (BPH), and some rare inherited metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text