Thermo Scientific Chemicals

Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%, Thermo Scientific Chemicals

Catalog number: 043646.22
100 g, Each
Thermo Scientific Chemicals

Calcium sulfide, 99% (metals basis excluding Mg), Mg <1%, Thermo Scientific Chemicals

Catalog number: 043646.22
100 g, Each
Quantity
Catalog number: 043646.22
also known as 043646-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20548-54-3
IUPAC Name
calcium sulfanediide
Molecular Formula
CaS
InChI Key
AGVJBLHVMNHENQ-UHFFFAOYSA-N
SMILES
[S--].[Ca++]
Impurity content
Mg <1%
Total Metal Impurities
1% max.

Description

Calcium sulfide is used in phosphors, lubricant additives and luminous paints. It reacts with elemental sulfur to give a lime-sulfur. Further, it is used in ore dressing and flotation agent. In addition to this, it is used as a depilatory in the tanning industry and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text