Thermo Scientific Chemicals

Copper(II) phthalocyanine, Thermo Scientific Chemicals

Catalog number: 043650.18
50 g, Each
Thermo Scientific Chemicals

Copper(II) phthalocyanine, Thermo Scientific Chemicals

Catalog number: 043650.18
50 g, Each
Quantity
Catalog number: 043650.18
also known as 043650-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147-14-8
IUPAC Name
9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Molecular Formula
C32H16CuN8
InChI Key
XCJYREBRNVKWGJ-UHFFFAOYSA-N
SMILES
[Cu]1N2C3=NC4=NC(=NC5=C6C=CC=CC6=C(N=C6N=C(N=C2C2=CC=CC=C32)C2=CC=CC=C62)N15)C1=CC=CC=C41
Assay (unspecified)
> 95.0% (dye content)
Appearance (Color)
Blue to purple
Form
powder

Description

Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text