Thermo Scientific Chemicals

Tetra-n-butylammonium hydroxide, 1.0M aq. soln., HPLC grade, Thermo Scientific Chemicals

Catalog number: 043662.AP
500 mL, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium hydroxide, 1.0M aq. soln., HPLC grade, Thermo Scientific Chemicals

Catalog number: 043662.AP
500 mL, Each
Quantity
Catalog number: 043662.AP
also known as 043662-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2052-49-5
IUPAC Name
tetrabutylazanium hydroxide
Molecular Formula
C16H37NO
InChI Key
VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
Optical Absorbance at
245 nm 0.10 max ( for 0.1 M solution)
Assay (Titration)
1.0 +/- 0.02 M
Optical Absorbance at
254 nm 0.07 max ( for 0.1 M solution)
Optical Absorbance at
300 nm 0.04 max ( for 0.1 M solution)
Optical Absorbance at
240 nm 0.15 max ( for 0.1 M solution)

Description

Phase transfer catalyst, electrolyte in polarography, ion-pair extractionTetra-n-butylammonium hydroxide, 1.0M in aqueous solution is used as a strong base in organic chemistry. It is utilized under phase-transfer conditions to effect alkylation and deprotonation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text