Thermo Scientific Chemicals

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

Catalog number: 043664.09
10 g, Each
Thermo Scientific Chemicals

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

Catalog number: 043664.09
10 g, Each
Quantity
Catalog number: 043664.09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12007-23-7
IUPAC Name
diboron hafnium
Molecular Formula
B2Hf
InChI Key
LRTTZMZPZHBOPO-UHFFFAOYSA-N
SMILES
[B].[B].[Hf]
Total Metal Impurities excluding Zr
0.5% max.
Impurity content
Zr<2%

Description

Hafnium boride is used in wear resistant coatings. It is also employed as a nuclear reactor control rod. Furthermore, it is used in hypervelocity re-entry vehicles like ICBM heat shields or aerodynamic leading-edges. It is useful in the preparation of high temperature composites in conjunction with silicon carbide (SiC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text