Thermo Scientific Chemicals
Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals
Catalog number: 043670.KG
1 Pc., Each
Thermo Scientific Chemicals
Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals
Catalog number: 043670.KG
1 Pc., Each
Quantity
Catalog number: 043670.KG
also known as 043670-KG
Price (USD)
Have Questions?
Description
Graphite rod, is suitable for making: Anodes, Electrodes, Heating elements, Crucibles, Molds & Ingots, Hot press dies, Gas nozzles, Lubrication sticks, Bushings & bearings.