Coal tar solution, contains 81.0-86.0% alcohol, Thermo Scientific Chemicals
Coal tar solution, contains 81.0-86.0% alcohol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coal tar solution, contains 81.0-86.0% alcohol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043671.K4
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
Add to cart
Coal tar solution, contains 81.0-86.0% alcohol, Thermo Scientific Chemicals
Catalog number043671.K4
Price (USD)/ Each
159.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
188.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS8007-45-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥81.0 to ≤86.0% alcohol
Appearance (Color)Yellow to dark green to dark brown
Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis). Coal tar has mild antiseptic and anti-inflammatory effects, and relieves itching. It is also absorbed into the skin cells and is thought to inhibit DNA replication in these cells. This slows down cell division and stops the cells from multiplying excessively. In turn, this reduces the thickening and scaling of the skin and clears up psoriasis plaques. It also finds its application in surface coatings and sealants. Also used as a fuel in open hearth furnaces in the steel industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use