Thermo Scientific Chemicals

Calcium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043674.22
100 g, Each
Thermo Scientific Chemicals

Calcium iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043674.22
100 g, Each
Quantity
Catalog number: 043674.22
also known as 043674-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-68-8
IUPAC Name
calcium diiodide
Molecular Formula
CaI2
InChI Key
UNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES
[Ca++].[I-].[I-]
Appearance (Color)
White
Residual water
≤100ppm
Form
Powder
Total Metal Impurities
≤0.01%

Description

Calcium iodide is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text