Polyethylene oxide, M.W. 1,000,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polyethylene oxide, M.W. 1,000,000, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 043678.36
also known as 043678-36
Price (USD)/ Each
410.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
483.00 
Save 72.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
410.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
483.00 
Save 72.35 (15%)
Add to cart
Polyethylene oxide, M.W. 1,000,000, Thermo Scientific Chemicals
Catalog number043678.36
Price (USD)/ Each
410.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
483.00 
Save 72.35 (15%)
-
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Viscosity400 to 800 cps ( 2% aq. solution, 25 deg C)
Polyethylene oxide is used as lubricants for rubber molds, textiles, metalworking and ceramics production. Further, it is used in food packaging, water-based paints, polishes, paper coatings, dispersing agents and chemical intermediates. In addition, it is used to impart viscosity to and modify the flow of aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only