Thermo Scientific Chemicals

Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043679.AE
100 mL, Each
Thermo Scientific Chemicals

Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043679.AE
100 mL, Each
Quantity
Catalog number: 043679.AE
also known as 043679-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-69-3
IUPAC Name
hexanedinitrile
Molecular Formula
C6H8N2
InChI Key
BTGRAWJCKBQKAO-UHFFFAOYSA-N
SMILES
N#CCCCCC#N
Form
Liquid
Appearance (Color)
Clear, colorless to pale yellow
Assay from Supplier's CofA
≥99.0%
Infrared spectrum
Conforms

Description

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text