Calcium molybdenum oxide, 99%, Thermo Scientific Chemicals
Calcium molybdenum oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium molybdenum oxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 043686.22
also known as 043686-22
Price (USD)/ Each
112.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
112.00
Add to cart
Calcium molybdenum oxide, 99%, Thermo Scientific Chemicals
Catalog number043686.22
Price (USD)/ Each
112.00
-
Add to cart
Chemical Identifiers
CAS7789-82-4
IUPAC Namecalcium dioxomolybdenumbis(olate)
Molecular FormulaCaMoO4
InChI KeyBIOOACNPATUQFW-UHFFFAOYSA-N
SMILES[Ca++].[O-][Mo]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only