Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals
Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043697.03
Price (USD)/ Each
127.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
127.00
Add to cart
Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals
Catalog number043697.03
Price (USD)/ Each
127.00
-
Add to cart
Chemical Identifiers
CAS13444-94-5
IUPAC Namepalladium(2+) dibromide
Molecular FormulaBr2Pd
InChI KeyINIOZDBICVTGEO-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Palladium(II) bromide is used as a source of the palladium(0) precatalyst in various coupling reactions. It is used as a catalyst for the carbonylative Heck coupling reaction. It is also used as an oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only