Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 Pk.
2 Pk.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043700.KV
also known as 043700-KV
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.60 
Save 10.95 (15%)
-
Add to cart
Quantity:
10 Pk.
Request bulk or custom format
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.60 
Save 10.95 (15%)
Add to cart
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Catalog number043700.KV
Price (USD)/ Each
63.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
74.60 
Save 10.95 (15%)
-
Add to cart
Chemical Identifiers
CAS7789-20-0
IUPAC Name[(²H)oxy](²H)
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES[2H]O[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMR> 99.95% isotopic enrichment
Deuterium oxide is used in nuclear magnetic resonance spectroscopy (NMR). It is also useful in the identification of labile hydrogens. As a source of deuterium, it is utilized for preparing specifically labeled isotopologs of organic compounds. It is often used instead of water for analyzing proteins in solution by using fourier transform infrared spectroscopy (FTIR). It finds application in certain types of nuclear reactors and in tritium production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only