Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043702.06
5 g, Each
Thermo Scientific Chemicals

Rhodium(III) chloride hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043702.06
5 g, Each
Quantity
Catalog number: 043702.06
also known as 043702-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20765-98-4
IUPAC Name
rhodium(3+) trichloride
Molecular Formula
Cl3Rh
InChI Key
SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Rh+3]
Total Metal Impurities
0.01% max.

Description

Rhodium(III) chloride hydrate is used as a catalyst for the conversion of methane to acetic acid, isomerization of alkenes, reduction of aromatic rings and hydration of acetylene. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals. It plays an important role as a catalyst for conjugate reduction of cinnamaldehydes followed by cross-coupling with arylboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text