Crystal Violet lactone
Crystal Violet lactone
Thermo Scientific Chemicals

Crystal Violet lactone

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043710.06
also known as 043710-06
Price (USD)/ Each
50.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
50.30
Add to cart
Crystal Violet lactone
Catalog number043710.06
Price (USD)/ Each
50.30
-
Add to cart
Chemical Identifiers
CAS1552-42-7
IUPAC Name6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC26H29N3O2
InChI KeyIPAJDLMMTVZVPP-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C1(OC(=O)C2=CC(=CC=C12)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
AbsorbancePale yellow to pale grey
FormPowder
Purity≥95.0%
Crystal Violet lactone is an important functional dye for the production of pressure sensitive materials. It is a chemical intermediate , intermediates for thermal/carbonless Paper, used in medicine and biochemical research, a leuco dye..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only