Thermo Scientific Chemicals

Crystal Violet lactone

Catalog number: 043710.14
25 g, Each
Thermo Scientific Chemicals

Crystal Violet lactone

Catalog number: 043710.14
25 g, Each
Quantity
Catalog number: 043710.14
also known as 043710-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1552-42-7
IUPAC Name
6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C26H29N3O2
InChI Key
IPAJDLMMTVZVPP-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C1(OC(=O)C2=CC(=CC=C12)N(C)C)C1=CC=C(C=C1)N(C)C
Absorbance
Pale yellow to pale grey
Form
Powder
Purity
≥95.0%

Description

Crystal Violet lactone is an important functional dye for the production of pressure sensitive materials. It is a chemical intermediate , intermediates for thermal/carbonless Paper, used in medicine and biochemical research, a leuco dye..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text