Thermo Scientific Chemicals

Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043712.36
500 g, Each
Thermo Scientific Chemicals

Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043712.36
500 g, Each
Quantity
Catalog number: 043712.36
also known as 043712-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2269-22-9
IUPAC Name
aluminium(3+) tris(butan-2-olate)
Molecular Formula
C12H27AlO3
InChI Key
WOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES
[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
Appearance (Color)
Colorless to pale yellow
Form
Viscous liquid
Assay from Supplier's CofA
≥94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text