Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043720.18
50 g, Each
Thermo Scientific Chemicals

Cesium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043720.18
50 g, Each
Quantity
Catalog number: 043720.18
also known as 043720-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-17-8
IUPAC Name
caesium(1+) chloride
Molecular Formula
ClCs
InChI Key
AIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES
[Cl-].[Cs+]
Appearance (Color)
White
Form
Beads
Residual water
≤100 ppm
Total Metal Impurities
≤0.01%

Description

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text