3-Hydroxy-3-methyl-2-butanone, 90+%, Thermo Scientific Chemicals
3-Hydroxy-3-methyl-2-butanone, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxy-3-methyl-2-butanone, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 043724.22
also known as 043724-22
Price (USD)/ Each
633.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
633.00
Add to cart
3-Hydroxy-3-methyl-2-butanone, 90+%, Thermo Scientific Chemicals
Catalog number043724.22
Price (USD)/ Each
633.00
-
Add to cart
Specifications
Chemical Name or Material3-Hydroxy-3-methyl-2-butanone
CAS115-22-0
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
3-Hydroxy-3-methyl-2-butanone used to produce 2,5-dihydroxy-2-methyl-heptan-3-one, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only