Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 1,300,000, Thermo Scientific Chemicals

Catalog number: 043728.A1
1 kg, Each
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 1,300,000, Thermo Scientific Chemicals

Catalog number: 043728.A1
1 kg, Each
Quantity
Catalog number: 043728.A1
also known as 043728-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-39-8
Molecular Formula
(C6H9NO)n
SMILES
*-CC(-*)N1CCCC1=O
MDL Number
MFCD01076626
Assay (unspecified)
K value (1% in water): 88-100
Appearance (Color)
White to off-white
Form
Powder
Assay (Titration)
Vinylpyrrolidone Monomer: ≤300 ppm
Water Content
≤5.0%

Description

Polyvinylpyrrolidone is used to make hair sprays, shampoos, plasma substitutes, detergents and adhesives. It is also used in hair dyes, clarifying agents in wine, beer, and juices, thickeners in inks and paints, water binding agents and protective colloids. It finds application as a special additive for batteries, ceramics, fiberglass, inks, and inkjet, an inhibitor of recrystallisation and to increase resolution in photoresists for cathode ray tubes. Further, it is used for gene delivery. In addition to this, it is used as a pharmaceutical aid, complexing agent and solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text