Thermo Scientific Chemicals

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: 043730.09
10 g, Each
Thermo Scientific Chemicals

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: 043730.09
10 g, Each
Quantity
Catalog number: 043730.09
also known as 043730-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
273-13-2
IUPAC Name
2,1,3-benzothiadiazole
Molecular Formula
C6H4N2S
InChI Key
PDQRQJVPEFGVRK-UHFFFAOYSA-N
SMILES
S1N=C2C=CC=CC2=N1
Form
Power or solid
Assay (GC)
> 97.5%
Appearance (Color)
White to off-white

Description

The product is used as an industrial solvent, and pharmaceutical intermediates for organic synthesis and used nitrocellulose and other resin, wax, fatty, dyes and other solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text