Thermo Scientific Chemicals

Antimony(III) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043732.22
100 g, Each
Thermo Scientific Chemicals

Antimony(III) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043732.22
100 g, Each
Quantity
Catalog number: 043732.22
also known as 043732-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-61-9
Residual water
< 100 ppm
Appearance (Color)
White
Form
Lump
Total Metal Impurities
0.001% max.
Particle Size
5-10mm

Description

Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text