Potassium heptafluorotantalate(V), 99.99% (metals basis), Thermo Scientific Chemicals
Potassium heptafluorotantalate(V), 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium heptafluorotantalate(V), 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 043733.18
also known as 043733-18
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
Potassium heptafluorotantalate(V), 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number043733.18
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS16924-00-8
IUPAC Namedipotassium heptafluorotantalumdiuide
Molecular FormulaF7H7K2Ta
InChI KeyBLXHHPZXEPALDX-UHFFFAOYSA-N
SMILESF.F.F.F.F.F.F.[K+].[K+].[Ta--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
It is an important raw material used as a primary and secondary pharmaceutical intermediate and also used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only