Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043741.5Y
4 x 100 mL, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 043741.5Y
4 x 100 mL, Each
Quantity
Catalog number: 043741.5Y
also known as 043741-5Y
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Form
Liquid
Appearance (Color)
Colorless to pale yellow
Color (APHA)
<30
Assay (GC)
≥99.5% (Corrected for water)
Comment
Inert gas used can be either Argon or Nitrogen

Description

1-methyl-2-pyrrolidinone is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility. It is used as stripping agent, oven cleaners and paint strippers. It is involved in the extraction of lube oil process and in the purification of synthetic gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text