Thermo Scientific Chemicals

Arsenic(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043748.06
5 g, Each
Thermo Scientific Chemicals

Arsenic(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043748.06
5 g, Each
Quantity
Catalog number: 043748.06
also known as 043748-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1303-36-2
Total Metal Impurities
0.001% max.

Description

Arsenicselenide is used as a vacuum deposition, chalcogenide glass for infrared optics, as it transmits light with wavelengths between 870 nm and 17.2 μm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text