Thermo Scientific Chemicals

Leucomalachite Green

Catalog number: 043758.04
2 g, Each
Thermo Scientific Chemicals

Leucomalachite Green

Catalog number: 043758.04
2 g, Each
Quantity
Catalog number: 043758.04
also known as 043758-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
129-73-7
IUPAC Name
4-{[4-(dimethylamino)phenyl](phenyl)methyl}-N,N-dimethylaniline
Molecular Formula
C23H26N2
InChI Key
WZKXBGJNNCGHIC-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
Appearance (Color)
White to pale green or pale blue
Form
Crystals or powder or crystalline powder
Comment
Material sourced in US and in other countries
Elemental Analysis (typical)
Carbon, C: 77.74-89.45% [US from vendor's CofA]
Elemental Analysis (typical)
Nitrogen, N: 7.88-9.07% [US from vendor's CofA]

Description

Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text