Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%, Thermo Scientific Chemicals
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 043760.A7
also known as 043760-A7
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
Add to cart
Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%, Thermo Scientific Chemicals
Catalog number043760.A7
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS16570-28-8
IUPAC Namedicopper(2+) (phosphonatooxy)phosphonate
Molecular FormulaCu2O7P2
InChI KeyPEVJCYPAFCUXEZ-UHFFFAOYSA-J
SMILES[Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Cu 33.5-35.5%
Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only