Sulfur powder, -325 mesh, 99.5%, Thermo Scientific Chemicals
Sulfur powder, -325 mesh, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur powder, -325 mesh, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 043766.36
also known as 043766-36
Price (USD)/ Each
60.65
Online exclusive
67.60 
Save 6.95 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.60 
Save 6.95 (10%)
Add to cart
Sulfur powder, -325 mesh, 99.5%, Thermo Scientific Chemicals
Catalog number043766.36
Price (USD)/ Each
60.65
Online exclusive
67.60 
Save 6.95 (10%)
-
Add to cart
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only