Thermo Scientific Chemicals

Polydimethylsiloxane, hydroxy terminated, M.W. 4200, Thermo Scientific Chemicals

Catalog number: 043769.AK
250 mL, Each
Thermo Scientific Chemicals

Polydimethylsiloxane, hydroxy terminated, M.W. 4200, Thermo Scientific Chemicals

Catalog number: 043769.AK
250 mL, Each
Quantity
Catalog number: 043769.AK
also known as 043769-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70131-67-8
MDL Number
MFCD01325012
Form
Clear liquid
Color (APHA)
100, max
Viscosity
80 to 130cSt

Description

Polydimethylsiloxane is frequently used in a liquid containing 4-5% silica gel, emulsifiers and preservatives. It is employed in applications like contact lenses, medical devices, elastomers, in shampoos, food (antifoaming agent), caulking, lubricants, kinetic sand , and heat-resistant tiles. PDMS is a common surfactant, component of defoamers and as an herbicidal penetran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text