Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 043771.K2
also known as 043771-K2
Price (USD)/ Each
32.65
Online exclusive
36.50 
Save 3.85 (11%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.50 
Save 3.85 (11%)
Add to cart
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Catalog number043771.K2
Price (USD)/ Each
32.65
Online exclusive
36.50 
Save 3.85 (11%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Titration)23.75-26.25% w/v
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only