Thermo Scientific Chemicals

Propargylamine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: 043775.03
1 g, Each
Thermo Scientific Chemicals

Propargylamine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: 043775.03
1 g, Each
Quantity
Catalog number: 043775.03
also known as 043775-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15430-52-1
Form
Yellow to cream
Infrared spectrum
Conforms
Assay from Supplier's CofA
≥94.0%
Form
Powder or crystals or crystalline powder

Description

Propargylamine hydrochloride is a useful intermediates in the preparation of drugs. It is used in pharmaceutical, industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text