Thermo Scientific Chemicals
Ethyl 2-cyanoacetoacetate, Thermo Scientific Chemicals
Catalog number: 043792.09
10 g, Each
Thermo Scientific Chemicals
Ethyl 2-cyanoacetoacetate, Thermo Scientific Chemicals
Catalog number: 043792.09
10 g, Each
Quantity
Catalog number: 043792.09
also known as 043792-09
Price (USD)
Quantity
-
Have Questions?
Description
It is used as a pharmaceutical intermediate. It is also used in the synthesis of fluorine-containing alkyl 2-cyano-3-oxocarboxylates and also in the syntheses of pyrazolo [1, 2-a] pyrazole and pyrazolo [5, 1-b] [1, 3] oxazine derivatives.