Thermo Scientific Chemicals

(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals

Catalog number: 043793.18
50 g, Each
Thermo Scientific Chemicals

(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals

Catalog number: 043793.18
50 g, Each
Quantity
Catalog number: 043793.18
also known as 043793-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-84-2
IUPAC Name
2-hydrazinylethan-1-ol
Molecular Formula
C2H8N2O
InChI Key
GBHCABUWWQUMAJ-UHFFFAOYSA-N
SMILES
NNCCO
Assay (GC)
> 90.0%
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow

Description

(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text