1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals
1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 043803.30
also known as 043803-30
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
Add to cart
1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals
Catalog number043803.30
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS822-06-0
IUPAC Name1,6-diisocyanatohexane
Molecular FormulaC8H12N2O2
InChI KeyRRAMGCGOFNQTLD-UHFFFAOYSA-N
SMILESO=C=NCCCCCCN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Colorless to pale yellow
FormLiquid
1,6-Diisocyanatohexane is used as a polymerizing agent in polyurethane paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only