Thermo Scientific Chemicals

Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043807.18
50 g, Each
Thermo Scientific Chemicals

Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 043807.18
50 g, Each
Quantity
Catalog number: 043807.18
also known as 043807-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1343-88-0
IUPAC Name
silicon(4+) magnesium(2+) trioxidandiide
Molecular Formula
MgO3Si
InChI Key
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)
100.39
Total Metal Impurities
1% max.

Description

Magnesium silicate is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, ant caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text