Thermo Scientific Chemicals
Iminodiacetic acid disodium salt hydrate, 97%, Thermo Scientific Chemicals
Catalog number: 043809.30
250 g, Each
Thermo Scientific Chemicals
Iminodiacetic acid disodium salt hydrate, 97%, Thermo Scientific Chemicals
Catalog number: 043809.30
250 g, Each
Quantity
Catalog number: 043809.30
also known as 043809-30
Price (USD)
Quantity
-
Have Questions?
Description
Sodium iminodiacetate dibasic hydrate (Iminodiacetic acid disodium salt monohydrate) has been used in the preparation of different iminodiacetic acid-agarose supports, useful for selective adsorption of large proteins.