Thermo Scientific Chemicals

Zirconium oxide, catalyst support, Thermo Scientific Chemicals

Catalog number: 043814.36
500 g, Each
Thermo Scientific Chemicals

Zirconium oxide, catalyst support, Thermo Scientific Chemicals

Catalog number: 043814.36
500 g, Each
Quantity
Catalog number: 043814.36
Price (USD)
Have Questions?
Surface Area
48 to 63 m2/g
Appearance (Color)
White to off-white
Assay (unspecified)
Total pore volume ( Hg intrusion porosimetry) : 0.23 cc/g min
Form
Pellets

Description

Catalyst carrier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text