Zirconium oxide, catalyst support, Thermo Scientific Chemicals
Zirconium oxide, catalyst support, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium oxide, catalyst support, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 043814.A4
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialZirconium oxide
Recommended StorageAmbient temperatures
Physical FormPellets
Quantity2.5 kg
EINECS Number215-227-2
View more
Catalyst carrier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only