Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal, Thermo Scientific Chemicals

Catalog number: 043832.A1
1 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal, Thermo Scientific Chemicals

Catalog number: 043832.A1
1 kg, Each
Quantity
Catalog number: 043832.A1
also known as 043832-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Appearance (Color)
White
Comment
Total pore volume, (Hg): 0.80 to 1.20 cc/g
Comment
Packing density: 325 to 475 kg/m³
Surface Area
220 to 280 m²/g
Form
Pellets

Description

Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text