Thermo Scientific Chemicals

Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals

Catalog number: 043833.A1
1 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals

Catalog number: 043833.A1
1 kg, Each
Quantity
Catalog number: 043833.A1
also known as 043833-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Specific Gravity
Report
Particle Size
OD Average: Report
Particle Size
Total Pore Volume: 0.45-6.0 ml/g
Surface Area
0.6-0.9 m²/g
Assay (unspecified)
Apparent Porosity: 60-70%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text