Thermo Scientific Chemicals
Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x1mm (0.394x0.394x0.039in), Thermo Scientific Chemicals
Catalog number: 043834.KF
1 Pc., Each
Thermo Scientific Chemicals
Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x1mm (0.394x0.394x0.039in), Thermo Scientific Chemicals
Catalog number: 043834.KF
1 Pc., Each
Quantity
Catalog number: 043834.KF
also known as 043834-KF
Price (USD)
Have Questions?
Comment
10x10x1mm
Comment
Thermo conductivity along C axis 6-10 watt/mK
Comment
Thermo conductivity along surface plane 1600-2000 watt/mK