5-Fluorosalicylaldehyde, 98+%, Thermo Scientific Chemicals
5-Fluorosalicylaldehyde, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluorosalicylaldehyde, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043844.03
also known as 043844-03
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
5-Fluorosalicylaldehyde, 98+%, Thermo Scientific Chemicals
Catalog number043844.03
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS347-54-6
IUPAC Name5-fluoro-2-hydroxybenzaldehyde
Molecular FormulaC7H5FO2
InChI KeyFDUBQNUDZOGOFE-UHFFFAOYSA-N
SMILESOC1=CC=C(F)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point82.0-86.0°C
Appearance (Color)White to yellow to orange to green
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA>98.0% (GC)
Assay from Supplier's CofA>98.0% (titration)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only