Thermo Scientific Chemicals

3,3'-Diethylthiadicarbocyanine iodide, Thermo Scientific Chemicals

Catalog number: 043851.04
2 g, Each
Thermo Scientific Chemicals

3,3'-Diethylthiadicarbocyanine iodide, Thermo Scientific Chemicals

Catalog number: 043851.04
2 g, Each
Quantity
Catalog number: 043851.04
also known as 043851-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
514-73-8
IUPAC Name
3-ethyl-2-[(3E)-5-[(2E)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular Formula
C23H23IN2S2
InChI Key
MNQDKWZEUULFPX-UHFFFAOYSA-M
SMILES
[I-].CCN1C(SC2=CC=CC=C12)=C/C=C/C=CC1=[N+](CC)C2=CC=CC=C2S1
lambda-max (UV-Vis)
650 to 660 nm
Appearance (Color)
Green
Form
powder`
Assay (unspecified)
absorptivity at lamda-max: > 175,000 L /mol.cm

Description

It is employed as a solvent soluble laser dye. DTDCi is used as fluorescent contrast agent in three-dimensional fluorescence lifetime tomography and as a dye for rapid genetic screening. It is a potential sensitive probe useful for fluorometric analysis of transferable membrane pores and effects of membrane potential on Na cotransports in eel intestinal brush-border membrane vesicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text