5,7-Dihydroxy-4'-methoxyisoflavone, 98%
5,7-Dihydroxy-4'-methoxyisoflavone, 98%
5,7-Dihydroxy-4'-methoxyisoflavone, 98%
5,7-Dihydroxy-4'-methoxyisoflavone, 98%
Thermo Scientific Chemicals

5,7-Dihydroxy-4'-methoxyisoflavone, 98%

CAS: 491-80-5 | C16H12O5 | 284.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 043853.MD
also known as 043853-MD
Price (USD)
65.65
Special offer
Online exclusive
Ends: 26-Sep-2025
81.50
Save 15.85 (19%)
Each
Estimated availability date 10-Jul-2025
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 26-Sep-2025
81.50
Save 15.85 (19%)
Each
Add to cart
Specifications
Beilstein278107
CAS491-80-5
ChEBICHEBI:17574
Chemical Name or Material5,7-Dihydroxy-4'-methoxyisoflavone
EINECS Number207-744-7
View more
Ask our AI about this Product
It has putative benefits in dietary cancer prophylaxis. It has also been found to inhibit fatty acid amide hydrolase and to act as agonist of PPARgamma, nuclear receptor that is current pharmacological target for the treatment of diabetes type 2. It acts as an antineoplastic agent. It is a selective agonist at ER-β estrogen receptors, and may have chemopreventive efficacy against breast cancer. In line with its low activity at ER-α estrogen receptors, it is essentially devoid of uterotrophic activity. Biochanin A is also a ligand for the aryl hydrocarbon receptor (AhR). It reduces arterial resistance and enhances microcirculation perhaps via effects on potassium and/or calcium ion channels. Induction of sulfotransferases for xenobiotic detoxification has been proposed as a mechanism of its cancer preventive effects. It is a nitric oxide synthase inhibitor and apoptosis inducer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
T02A059Certificate of AnalysisApr 12, 2022043853.03, 043853.06, 043853.MD
C23X034Certificate of AnalysisDec 07, 2021043853.03, 043853.06, 043853.MD
J16S041Certificate of AnalysisNov 29, 2021043853.03, 043853.06, 043853.MD
B27S028Certificate of AnalysisNov 23, 2021043853.03, 043853.06, 043853.MD
G29P24Certificate of AnalysisNov 23, 2021043853.03, 043853.06, 043853.MD
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection