Thermo Scientific Chemicals

N,N-Dimethylacetamide-d{9}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog number: 043854.03
1 g, Each
Thermo Scientific Chemicals

N,N-Dimethylacetamide-d{9}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog number: 043854.03
1 g, Each
Quantity
Catalog number: 043854.03
also known as 043854-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
116057-81-9
Assay from Supplier's CofA
≥99.0% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA
≥98.0% ¹H nmr
Water Content (Karl Fischer Titration)
≤ 500ppm
Appearance (Color)
Clear colorless
Form
Liquid

Description

N,N-Dimethylacetamide-d{9} is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text