Thermo Scientific Chemicals

Aluminum oxide, catalyst support, intermediate surface area (low SiO{2}), Thermo Scientific Chemicals

Catalog number: 043857.A7
5 kg, Each
Thermo Scientific Chemicals

Aluminum oxide, catalyst support, intermediate surface area (low SiO{2}), Thermo Scientific Chemicals

Catalog number: 043857.A7
5 kg, Each
Quantity
Catalog number: 043857.A7
also known as 043857-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Appearance (Color)
White to pale cream
Surface Area
80-120 m²/g
Form
Pellets
Comment
Total pore volume: 0.44-0.54 cc/g

Description

Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text