Platinum, nominally 10% on carbon black, HiSPEC™ 2000, Thermo Scientific Chemicals
Platinum, nominally 10% on carbon black, HiSPEC™ 2000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, nominally 10% on carbon black, HiSPEC™ 2000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
0.5 g
10 g
Catalog number 043876.04
also known as 043876-04
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
Add to cart
Platinum, nominally 10% on carbon black, HiSPEC™ 2000, Thermo Scientific Chemicals
Catalog number043876.04
Price (USD)/ Each
173.65
Online exclusive
193.00 
Save 19.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum
Name NoteNominally 10% on carbon black, HiSPEC™ 2000
CAS7440-6-6
Health Hazard 1H351
Health Hazard 2GHS H Statement
H228
Flammable solid.
View more
Platinum black commonly acts as a catalyst in chemical reactions, as a catalytic converter in automobiles and in the petroleum industry as a catalyst. It also vigorously catalyzes the decomposition of hydrogen peroxide into water and oxygen. It is used extensively as laboratory equipment, electrical contacts,electrodes, platinum resistance thermometers, dentistry equipment, and jewellery. Compounds containing platinum (cisplatin, oxaliplatin and carboplatin) are applied in chemotherapy against certain types of cancer and also applied as a pollution control catalysts for gasoline and diesel vehicles and in control of emissions of organics and nitrous oxides from industrial installations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only