Thermo Scientific Chemicals

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043879.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 043879.06
5 g, Each
Quantity
Catalog number: 043879.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-90-3
IUPAC Name
gallium(3+) trichloride
Molecular Formula
Cl3Ga
InChI Key
UPWPDUACHOATKO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ga+3]
Appearance (Color)
White to black
Form
Crystalline
Residual water
≤100 ppm
Total Metal Impurities
0.001% max.

Description

Gallium(III) chloride is used as lewis acid or precursor of organogallium reagents. It is used in detection of solar neutrinos. It is used in semiconductor devices for microelectronics and optics. It acts as a catalyst for a number of transformations such as the ring opening and recyclization of epoxide or trimerization of alkynes and as diamagnetic analogue for ferric compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text