Thermo Scientific Chemicals

Glycerol monostearate, purified, Thermo Scientific Chemicals

Catalog number: 043883.30
250 g, Each
Thermo Scientific Chemicals

Glycerol monostearate, purified, Thermo Scientific Chemicals

Catalog number: 043883.30
250 g, Each
Quantity
Catalog number: 043883.30
also known as 043883-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31566-31-1
IUPAC Name
2,3-dihydroxypropyl octadecanoate
Molecular Formula
C21H42O4
InChI Key
VBICKXHEKHSIBG-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥95.0%
Comment
Product is a mixture of monosterate, monopalmitate and diglyceride (<10%)
Iodine value
≤6.0
Form
Powder

Description

Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text